Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152158 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152158 Supplier Novus Biologicals Supplier No. NBP152158
Add to Cart
Add to Cart

Goat Polyclonal Antibody

FBXO4 Polyclonal specifically detects FBXO4 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen FBXO4
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_036308.1
Gene Alias DKFZp547N213, F-box protein 4, F-box protein Fbx4, FBX4F-box only protein 4, FLJ10141
Gene Symbols FBXO4
Host Species Goat
Immunogen Peptide with sequence C-EHTSAVNKMFSRH corresponding to internal region according to NP_277019.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26272
Test Specificity This antibody is expected to recognize isoform 1 and 2 (NP_036308.1; NP_277019.1).
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.