Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO44 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB345143 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345143 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345143 Supplier Novus Biologicals Supplier No. NB100467F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBXO44 Polyclonal specifically detects FBXO44 in Human samples. It is validated for Western Blot.

Specifications

Antigen FBXO44
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias FBG3 DKFZp781J0852, F-box gene 3, F-box only protein 44, F-box protein 44, F-box protein FBX30, F-box/G-domain protein 3, FBX30FBX6AFbx44, FBX44, Fbxo6a, MGC14140
Host Species Rabbit
Immunogen A synthetic peptide containing residues 103-119 EWKVEDLSRDQRKEFPN) of the human FBG3 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93611
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.