Learn More
Description
Specifications
Specifications
| Antigen | FBXW11 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q9UKB1 |
| Gene Alias | beta-transducin repeat-containing protein 2, BTRC2F-box/WD repeat-containing protein 1B, BTRCP2FBW1B, F-box and WD repeat domain containing 11, F-box and WD-40 domain protein 11, F-box and WD-40 domain protein 1B, F-box protein Fbw1b, F-box/WD repeat-containing protein 11, Fbw11, FBXW1BFbw1b, Homologous to Slimb protein, Hos, KIAA0696F-box and WD repeats protein beta-TrCP2 |
| Gene Symbols | FBXW11 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
