Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXW7/Cdc4 Antibody (FBOX 3a9/1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP50403549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP50403549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP50403549 Supplier Novus Biologicals Supplier No. NBP250403JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FBXW7/Cdc4 Monoclonal specifically detects FBXW7/Cdc4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen FBXW7/Cdc4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone FBOX 3a9/1
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AGOFBXO30, Archipelago homolog, archipelago, Drosophila, homolog of, CDC4FLJ16457, F-box and WD repeat domain containing 7, F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila), F-box and WD-40 domain-containing protein 7, F-box protein FBW7, F-box protein FBX30, F-box protein SEL-10, F-box/WD repeat-containing protein 7, FBW7hCdc4, FBX30FLJ11071, FBXW6, hAgo, homolog of C elegans sel-10, SEL-10DKFZp686F23254, SEL10FBW6
Gene Symbols FBXW7
Host Species Mouse
Immunogen Synthetic peptide of human sequence
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 55294
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.