Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXW7/Cdc4 Antibody (OTI6F5), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N271995A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271995A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271995A700 Supplier Novus Biologicals Supplier No. NBP271995AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FBXW7/Cdc4 Monoclonal specifically detects FBXW7/Cdc4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FBXW7/Cdc4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6F5
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AGOFBXO30, Archipelago homolog, archipelago, Drosophila, homolog of, CDC4FLJ16457, F-box and WD repeat domain containing 7, F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila), F-box and WD-40 domain-containing protein 7, F-box protein FBW7, F-box protein FBX30, F-box protein SEL-10, F-box/WD repeat-containing protein 7, FBW7hCdc4, FBX30FLJ11071, FBXW6, hAgo, homolog of C elegans sel-10, SEL-10DKFZp686F23254, SEL10FBW6
Gene Symbols FBXW7
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-354 of human FBXW7/Cdc4 (NP_361014) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 55294.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.