Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXW9 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB330903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330903 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330903 Supplier Novus Biologicals Supplier No. NBP297608MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBXW9 Polyclonal antibody specifically detects FBXW9 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen FBXW9
Applications Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias F-box and WD repeat domain containing 9, F-box and WD-40 domain protein 9, F-box and WD-40 domain-containing protein 9, F-box and WD-40 repeat containing protein 9, specificity factor for SCF ubiquitin ligase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FBXW9 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84261
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.