Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc epsilon RI alpha Antibody (773735), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB302917 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302917 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302917 Supplier Novus Biologicals Supplier No. FAB66781Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc epsilon RI alpha Monoclonal antibody specifically detects Fc epsilon RI alpha in Human samples. It is validated for Serum

Specifications

Antigen Fc epsilon RI alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 773735
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias FCE1A,fcepsilonri,FCER1A,FcERI,Fcr-5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fc epsilon RI alpha , Val26-Gln205, Accession # P12319
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.