Learn More
Description
Specifications
Specifications
| Antigen | Fc epsilon RI |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | AER-37 |
| Conjugate | DyLight 680 |
| Dilution | Flow Cytometry, Immunohistochemistry-Paraffin |
| Gene Alias | alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide |
| Gene Symbols | FCER1A |
| Host Species | Mouse |
| Immunogen | The immunogen for this antibody was FceRI alpha. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
