Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc epsilon RI Antibody (IE7), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB154001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154001 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB154001 Supplier Novus Biologicals Supplier No. NBP312052DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen Fc epsilon RI
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone IE7
Conjugate DyLight 594
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide
Host Species Rabbit
Immunogen Recombinant human protein, N-terminus.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2205
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.