Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RI/CD64 Antibody (008), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB333327 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333327 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333327 Supplier Novus Biologicals Supplier No. NBP290404MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Fc gamma RI/CD64
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 008
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Fc gamma RI/CD64 (Accession#: NP_034316.1, Met 1-Pro 297)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2209
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.