Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RI/CD64 Antibody (32.2-1A), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB314667 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314667 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314667 Supplier Novus Biologicals Supplier No. FAB12573PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Fc gamma RI/CD64
Applications Flow Cytometry
Classification Monoclonal
Clone 32.2-1A
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Host Species Mouse
Immunogen Partially purified detergent lysate of the high affinity Fc receptor from U937 cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2209
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.