Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RI/CD64 Antibody (H22), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB016000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016000 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016000 Supplier Novus Biologicals Supplier No. NBP281097FR
Add to Cart
Add to Cart

Human Monoclonal Antibody

Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Competitive ELISA.

Specifications

Antigen Fc gamma RI/CD64
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA
Classification Monoclonal
Clone H22
Conjugate DyLight 680
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence, Competitive ELISA
Formulation 50mM Sodium Borate
Gene Alias CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Gene Symbols FCGR1A
Host Species Human
Immunogen This recombinant antibody was prepared by cloning the single chain fragment (scFv) derived from the murine anti-human Fc gamma RI/CD64 mAb m22 into the bacterial expression vector pBM1.1, and fusing it to a deletion mutant of Pseudomonas exotoxin A.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2209
Test Specificity This immunotoxin is specific for human Fc gamma RI/CD64, and specifically an epitope outside the Fc binding domain. The binding activity of the VH/VL antibody format was unaffected by fusion of the m22(scFv) coding regions to the truncated ETA coding sequences
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.