Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RII/CD32 Antibody (190723) [Alexa Fluor™ 532], Novus Biologicals™
SDP

Catalog No. NB400350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400350 Supplier Novus Biologicals Supplier No. FAB1330X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen Fc gamma RII/CD32
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 190723
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIB, Ala46-Pro217, Accession # P31994
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2213
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.