Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RII/CD32 Antibody (FCGR2A/479), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB382705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382705 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382705 Supplier Novus Biologicals Supplier No. NBP308220AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Fc gamma RII/CD32
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone FCGR2A/479
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Host Species Mouse
Immunogen Recombinant human Fc gamma RII/CD32 protein (Uniprot: P12318)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2213
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.