Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RII/RIII (CD32/CD16) Antibody (2.4G2) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25264402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25264402 0.2 mg
NBP25264425 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25264402 Supplier Novus Biologicals Supplier No. NBP2526440.2MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD16/CD32 Monoclonal specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, CyTOF-ready, Block/Neutralize.

Specifications

Antigen CD16/CD32
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Blocking Assay, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 2.4G2
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Gene Symbols FCGR3A
Host Species Rabbit
Immunogen Mouse BALB/c Macrophage J774 Cell Line.
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity This antibody binds to an epitope on the extracellular domains of the mouse FcyIII and FcyII, thereby inhibiting their ability to bind antibodies via their Fc region.receptors. Also binding the FcyI receptor (CD64) via its Fc domain.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.