Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RII/RIII (CD32/CD16) Antibody (2.4G2) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$268.00 - $584.50

Specifications

Antigen CD16/CD32
Clone 2.4G2
Dilution Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Blocking Assay, Immunofluorescence, CyTOF
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25264425
SDP
View Documents
Novus Biologicals
NBP2526440.025MG
0.025 mg
Each for $268.00
Only null left
Add to Cart
 
NBP25264402
SDP
View Documents
Novus Biologicals
NBP2526440.2MG
0.2 mg
Each for $584.50
Only null left
Add to Cart
 
Description

Description

CD16/CD32 Monoclonal specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, CyTOF-ready, Block/Neutralize.
Specifications

Specifications

CD16/CD32
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready
Monoclonal
Purified
RUO
PBS with 0.02% Proclin 300. with 0.02% Proclin 300
2214
Mouse BALB/c Macrophage J774 Cell Line.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
2.4G2
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Blocking Assay, Immunofluorescence, CyTOF
Unconjugated
Rabbit
Mouse
CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
FCGR3A
IgG
Protein A or G purified
This antibody binds to an epitope on the extracellular domains of the mouse FcyIII and FcyII, thereby inhibiting their ability to bind antibodies via their Fc region.receptors. Also binding the FcyI receptor (CD64) via its Fc domain.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.