Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RII/RIII (CD32/CD16) Antibody (2.4G2), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB343239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343239 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343239 Supplier Novus Biologicals Supplier No. NBP252644DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Fc gamma RII/RIII (CD32/CD16) Monoclonal antibody specifically detects Fc gamma RII/RIII (CD32/CD16) in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF, Neutralization

Specifications

Antigen Fc gamma RII/RIII (CD32/CD16)
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone 2.4G2
Conjugate DyLight 594
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF-ready, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias CD16;CD16A;CD16-II;FCG3;FCGR3;FCGR3A;FCGRIII;FcGRIIIA;FCR-10;FCRIII;FCRIIIA;IGFR3;IMD20;Low affinity immunoglobulin gamma Fc region receptor III-A
Host Species Rabbit
Immunogen Mouse BALB/c Macrophage J774 Cell Line.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2214
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.