Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIA/CD32a Antibody (020), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB107725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107725 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107725 Supplier Novus Biologicals Supplier No. NBP289456DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Fc gamma RIIA/CD32a Monoclonal specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Western Blot.

Specifications

Antigen Fc gamma RIIA/CD32a
Applications Western Blot
Classification Monoclonal
Clone 20
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Fc gamma RIIA/CD32a (AAA35827.1; Met1-Ile218).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2212
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.