Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIA/CD32a Antibody (3L1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP235936 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP235936 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP235936 Supplier Novus Biologicals Supplier No. NBP235936

Mouse Monoclonal Antibody

Fc gamma RIIA/CD32a Monoclonal specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen Fc gamma RIIA/CD32a
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 3L1
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready
Gene Accession No. P31994
Gene Alias CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a
Gene Symbols FCGR2A
Host Species Mouse
Immunogen Human recombinant CD32 EC domain
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2212
Reconstitution Centrifuge vial prior to opening. Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.