Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIA/CD32a Antibody (OTI9G5), Novus Biologicals™
SDP

Catalog No. NBP147735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147735 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147735 Supplier Novus Biologicals Supplier No. NBP147735
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc gamma RIIA/CD32a Monoclonal specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Fc gamma RIIA/CD32a
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI9G5
Concentration 0.66 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 2ug/500ul, Immunohistochemistry-Paraffin 1:50
Gene Alias CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a
Gene Symbols FCGR2A
Host Species Mouse
Immunogen Full length human recombinant protein of human FCGR2A (NP_067674) produced in HEK293T cell.
Molecular Weight of Antigen 34.7 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2212
Test Specificity This antibody is specific for Homo sapiens Fc fragment of IgG, low affinity IIa, receptor (CD32) (FCGR2A), transcript variant 2.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.