Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIB/CD32b Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB326935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326935 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326935 Supplier Novus Biologicals Supplier No. NBP299136MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fc gamma RIIB/CD32b Polyclonal antibody specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for ELISA

Specifications

Antigen Fc gamma RIIB/CD32b
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Fc gamma RIIB/CD32b (Accession#: Ala 46-Pro 217; NP_001002274.1)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2213
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.