Learn More
Description
Specifications
Specifications
| Antigen | Fc gamma RIIB/CD32b |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 112 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b |
| Host Species | Rabbit |
| Immunogen | Recombinant Human Fc gamma RIIB/CD32b protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
