Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIB/CD32b Rabbit anti-Human, Clone: 112, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
| Antigen | Fc gamma RIIB/CD32b |
|---|---|
| Clone | 112 |
| Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Fc gamma RIIB/CD32b Monoclonal specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
| Fc gamma RIIB/CD32b | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| Recombinant Human Fc gamma RIIB/CD32b protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 112 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology, Phospho Specific | |
| 0.2 um filtered solution in PBS | |
| 2213 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title