Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIII (CD16) Antibody (015), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB332830 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332830 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332830 Supplier Novus Biologicals Supplier No. NBP290536MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Fc gamma RIII (CD16) Monoclonal antibody specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Fc gamma RIII (CD16)
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 015
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Fc gamma RIII (CD16) (Uniprot#: P08508-1, Met 1-Thr 215)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2214
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.