Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIII (CD16) Antibody (1001049), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB370312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370312 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370312 Supplier Novus Biologicals Supplier No. FAB4325Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.

Specifications

Antigen Fc gamma RIII (CD16)
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1001049
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human Fc gamma RIIIA/CD16a, Gly17-Gln208, Accession # P08637
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2214
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.