Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIII (CD16) Antibody (3G8) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119725 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119725 Supplier Novus Biologicals Supplier No. NBP309102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD16/CD32 Monoclonal specifically detects CD16/CD32 in Human, Primate samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional.

Specifications

Antigen CD16/CD32
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone 3G8
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 6 ug/ml, Immunoprecipitation, Immunohistochemistry-Frozen, Functional
Formulation PBS, 0.2 um filter sterilized
Gene Alias CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Host Species Mouse
Immunogen Human neutrophils
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2214
Target Species Human, Primate
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.