Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIII (CD16) Antibody (5B11), FITC, Novus Biologicals™
SDP

Catalog No. NBP242228F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242228F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242228F Supplier Novus Biologicals Supplier No. NBP242228F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Fc gamma RIII (CD16)
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5B11
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
Gene Symbols FCGR3A
Host Species Mouse
Immunogen Raised against CD16 antigen
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity Recognizes CD16 antigen
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.