Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIIA/CD16a Antibody (2829B), PE, Novus Biologicals™
SDP

Catalog No. NB305510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305510 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305510 Supplier Novus Biologicals Supplier No. FAB107512P

Rabbit Monoclonal Antibody

Fc gamma RIIIA/CD16a Monoclonal antibody specifically detects Fc gamma RIIIA/CD16a in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen Fc gamma RIIIA/CD16a
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2829B
Conjugate PE
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Formulation PBS
Gene Alias CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived human Fc gamma RIIIA/CD16a, Gly17-Gln208, Accession # P08637
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2214
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.