Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIIA/CD16a Antibody (3G8), PE, Novus Biologicals™
SDP

Catalog No. NB016132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016132 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016132 Supplier Novus Biologicals Supplier No. NBP281102PE

Rabbit Monoclonal Antibody

Fc gamma RIIIA/CD16a Monoclonal specifically detects Fc gamma RIIIA/CD16a in Human, Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Product Image.

Specifications

Antigen Fc gamma RIIIA/CD16a
Applications Flow Cytometry
Classification Monoclonal
Clone 3G8
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA
Gene Symbols FCGR3A
Host Species Rabbit
Immunogen 3G8 was prepared by immunizing mice with human PMN (polymorphonuclear leukocytes) cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity 3G8 binds to CD16 at an epitope which is not the IgG-binding site. This antibody cannot distinguish between CD16a transmembrane isoform found on the surface of NK cells and macrophages (which can trigger the cytolytic response), and the GPI-linked CD16b isoform present on neutrophils or soluble CD16 found in the plasma. CD16 is the low affinity IgG receptor III and binds to aggregated IgG or IgG-antigen complex which functions in NK cell activation, phagocytosis and ADCC (antibody-dependent cell-mediated cytotoxicity).
Target Species Human, Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus Macaque
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.