Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIIA/CD16a Antibody (DJ130c), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10064346G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064346G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064346G Supplier Novus Biologicals Supplier No. NB10064346G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc gamma RIIIA/CD16a Monoclonal specifically detects Fc gamma RIIIA/CD16a in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen Fc gamma RIIIA/CD16a
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DJ130c
Conjugate DyLight 488
Dilution Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA
Gene Symbols FCGR3A
Host Species Mouse
Immunogen CD34 antigen
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity NB100-64346 recognizes human CD16, a 50-65kD cell surface molecule, which is the low affinity receptor for IgG (FcR III). CD16 exists as a transmembranous form (Fc gammaRIIIA, or CD16A) and a glycosyl phosphatidylinositol (GPI) anchored form (Fc gammaRIIIB, or CD16B). CD16A is expressed by NK cells, some T cells, and macrophages, whereas CD16B is primarily expressed by granulocytes. In addition, CD16B exists as two allelic variants NA1 and NA2. DJ130c recognizes all polymorphonuclear cells irrespective on their NA phenotype. Studies show that the DJ130c antibody recognizes an epitope in the first membrane-distal domain of CD16 and recognizes both CD16A and CD16B.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.