Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIIA/CD16a Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB052736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052736 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052736 Supplier Novus Biologicals Supplier No. NBP298147DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fc gamma RIIIA/CD16a Polyclonal specifically detects Fc gamma RIIIA/CD16a in Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Fc gamma RIIIA/CD16a
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Fc gamma RIIIA/CD16a (ABO70343.1; Met 1-Pro 200).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2214
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.