Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIIB/CD16b Antibody (24), HRP, Novus Biologicals™
SDP

Catalog No. NB388683 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388683 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388683 Supplier Novus Biologicals Supplier No. NBP312795H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fc gamma RIIIB/CD16b Monoclonal antibody specifically detects Fc gamma RIIIB/CD16b in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Fc gamma RIIIB/CD16b
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 24
Conjugate HRP
Formulation PBS
Gene Alias CD16, CD16b, CD16b antigen, Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc fragment of IgG, low affinity IIIb, receptor for (CD16), FCG3, Fc-gamma receptor IIIb (CD 16), Fc-gamma RIII, Fc-gamma RIIIb, Fc-gamma RIII-beta, FCGR3, FcR-10, fcRIII, fcRIIIb, IGFR3, IgG Fc receptor III-1, low affinity immunoglobulin gamma Fc region receptor III-B
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Fc gamma RIIIB/CD16b (Uniprot#: O75015.2; Met1-Ser200).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2215
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.