Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIIB/CD16b Antibody (2830D), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB297076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297076 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297076 Supplier Novus Biologicals Supplier No. FAB15971E

Rabbit Monoclonal Antibody

Fc gamma RIIIB/CD16b Monoclonal antibody specifically detects Fc gamma RIIIB/CD16b in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Fc gamma RIIIB/CD16b
Applications Flow Cytometry
Classification Monoclonal
Clone 2830D
Conjugate DyLight 405
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD16, CD16b, CD16b antigen, Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc fragment of IgG, low affinity IIIb, receptor for (CD16), FCG3, Fc-gamma receptor IIIb (CD 16), Fc-gamma RIII, Fc-gamma RIIIb, Fc-gamma RIII-beta, FCGR3, FcR-10, fcRIII, fcRIIIb, IGFR3, IgG Fc receptor III-1, low affinity immunoglobulin gamma Fc region receptor III-B
Host Species Rabbit
Immunogen HEK293-derived human Fc gamma RIIIB/CD16b protein, Met18-Ser200, Accession # NP_000561
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2215
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.