Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIIB/CD16b Mouse anti-Human, Alexa Fluor 405, Clone: MM0272-5L11, Novus Biologicals™
SDP

Catalog No. NB212148405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212148405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212148405 Supplier Novus Biologicals Supplier No. NBP212148AF405

Mouse Monoclonal Antibody

Fc gamma RIIIB/CD16b Monoclonal antibody specifically detects Fc gamma RIIIB/CD16b in Human samples. It is validated for Western Blot.

Specifications

Antigen Fc gamma RIIIB/CD16b
Applications Western Blot
Classification Monoclonal
Clone MM0272-5L11
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD16, CD16b, CD16b antigen, Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc fragment of IgG, low affinity IIIb, receptor for (CD16), FCG3, Fc-gamma receptor IIIb (CD 16), Fc-gamma RIII, Fc-gamma RIIIb, Fc-gamma RIII-beta, FCGR3, FcR-10, fcRIII, fcRIIIb, IGFR3, IgG Fc receptor III-1, low affinity immunoglobulin gamma Fc region receptor III-B
Gene Symbols FCGR3B
Host Species Mouse
Immunogen Human CD16b
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2215
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.