Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCAR/CD89 Antibody (488032), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB371184 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371184 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371184 Supplier Novus Biologicals Supplier No. FAB3939J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Cynomolgus Monkey, Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen FCAR/CD89
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 488032
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human FCAR/CD89, Gln22-Lys287, Accession # P24071
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2204
Target Species Cynomolgus Monkey, Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.