Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCAR/CD89 Antibody (MIP8a), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N106326P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106326P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106326P594 Supplier Novus Biologicals Supplier No. NB10063266PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Human samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen FCAR/CD89
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MIP8a
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Gene Symbols FCAR
Host Species Mouse
Immunogen Recombinant soluble human Fc alpha R
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2204.0
Test Specificity NB100-63266 recognizes the human CD89 cell surface antigen, a 50-75kD cell surface glycoprotein that is also known as the IgA receptor (Fc alpha R). CD89 is expressed by peripheral blood monocytes and neutrophils. Clone MIP8a blocks binding of IgA to the Fc alpha R, and also inhibits neutrophil phagocytosis of IgA complexes.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.