Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FCCP
GREENER_CHOICE

Catalog No. p-4783453
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Oxidative phosphorylation uncoupler

A very potent uncoupler of oxidative phosphorylation in mitochondria.

Chemical Identifiers

CAS 370-86-5
Molecular Formula C10H5F3N4O

Specifications

Quantity 10 mg
Formula Weight 254.17
Purity >99%
Molecular Weight (g/mol) 254.172
Chemical Name or Material FCCP
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.