Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc epsilon RI alpha Mouse anti-Human, Clone: AH24-32, Novus Biologicals™
SDP

Catalog No. NBP267276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267276 Supplier Novus Biologicals Supplier No. NBP267276

Mouse Monoclonal Antibody

Fc epsilon RI alpha Monoclonal specifically detects Fc epsilon RI alpha in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FcepsilonRIalpha
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AH24-32
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:10000-1:50000, Immunohistochemistry 1:50-1:200 Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 1% BSA, 40% Glycerol with 0.2% Sodium Azide
Host Species Mouse
Immunogen Full length protein
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.