Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR2C Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096252 20 μL
NB096251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096252 Supplier Novus Biologicals Supplier No. NBP30503220UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FCGR2C Polyclonal specifically detects FCGR2C in Human samples. It is validated for Western Blot.

Specifications

Antigen FCGR2C
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias low affinity immunoglobulin gamma Fc region receptor II-c, CD32, CD32C, CDW32, Fc fragment of IgG, low affinity IIc, receptor for (CD32) (gene/pseudogene), FCG2, FCRIIC, IGFR2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 43-223 of human FCGR2C (NP_963857.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9103
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.