Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCHO2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP236524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236524 Supplier Novus Biologicals Supplier No. NBP236524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FCHO2 Polyclonal specifically detects FCHO2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen FCHO2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_620137.2
Gene Alias FCH domain only 2
Gene Symbols FCHO2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 125 to 175 of human FCH Domain Only 2 using the numbering given in entry NP_620137.2
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115548
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.