Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCHSD1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241137 0.1 mg
NB396489 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241137 Supplier Novus Biologicals Supplier No. NBP241137
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

FCHSD1 Polyclonal specifically detects FCHSD1 in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen FCHSD1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, ELISA
Formulation PBS with 0.02% Sodium Azide
Gene Alias FCH and double SH3 domains 1, FCH and double SH3 domains protein 1, FLJ00007, nervous wreck homolog 2, NWK2
Gene Symbols FCHSD1
Host Species Chicken
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human FCHSD1. The immunogen is located within the last 50 amino acids of FCHSD1. Amino Acid Squence: PPAKAPDPGHPDPLT
Molecular Weight of Antigen 82 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89848
Test Specificity Multiple isoforms of FCHSD1 are known to exist. FCHSD1 antibody is predicted to not cross-react with FCHSD2
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.