Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FcRH5, Mouse anti-Human, Clone: F25, Millipore Sigma™
Mouse Monoclonal Antibody
$207.00 - $570.00
Specifications
| Antigen | FcRH5 |
|---|---|
| Clone | F25 |
| Dilution | Flow Cytometry Analysis: A representative lot detected FcRH5 in Flow Cytometry applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).Western Blotting Analysis: A representative lot detected FcRH5 in Western Blotting applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).ELISA Analysis: A representative lot detected FcRH5 in ELISA applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96). |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF210325
|
MilliporeSigma
MABF210325UG |
25 μg |
Each for $207.00
|
|
|||||
|
MABF2103100
|
MilliporeSigma
MABF2103100UG |
100 μg |
Each for $570.00
|
|
|||||
Description
Anti-FcRH5, clone F25, Cat. No. MABF2103, is a mouse monoclonal antibody that detects Fc receptor-like protein 5 and has been tested for use in ELISA, Flow Cytometry, and Western Blotting.
Fc receptor-like protein 5 (UniProt: Q96RD9; also known as FcR-like protein 5, FcRL5, BXMAS1, Fc receptor homolog 5, FcRH5, Immune receptor translocation-associated protein 2, IRTA2, CD307e) is encoded by the FCRL5 (also known as FCRH5, IRTA2, UNQ503/PRO820) gene (Gene ID: 83416) in human. FcRH5 is a single-pass type I membrane protein that is expressed in marginal zone B-cells, immunoblasts, tonsillar germinal center centrocytes and in the intraepithelial and interfollicular regions of the tonsil. It is involved in B-cell development and differentiation in peripheral lymphoid organs and may be useful markers of B-cell stages. It is shown to regulate BCR signaling and binds aggregated IgG and engages several of its Ig-domains to bind IgG. It is shown to be overexpressed in many lymphoma cell lines and on hairy cell leukemia cells. Serum levels of soluble FcRH5 are reported to be elevated in patients with several types of B cell tumors. FcRH5 is synthesized with a signal peptide (aa 1-15), which is cleaved off in the mature form that has a long extracellular domain (aa 16-851), a short transmembrane domain (aa 852-872), and a cytoplasmic tail (aa 873-977) that contains immunoreceptor tyrosine-based inhibitor motifs (ITIM). (Ref.: Franco, A., et al. (2013). J. Immunol. 190(11); 5739-46; Ise, T., et al. (2007). Leukemia 21(1); 169-174).Specifications
| FcRH5 | |
| Flow Cytometry Analysis: A representative lot detected FcRH5 in Flow Cytometry applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).Western Blotting Analysis: A representative lot detected FcRH5 in Western Blotting applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).ELISA Analysis: A representative lot detected FcRH5 in ELISA applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96). | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Fc receptor-like protein 5;FcR-like protein 5;FcRL5;BXMAS1;Fc receptor homolog 5;Immune receptor translocation-associated protein 2;IRTA2;CD307e | |
| FCRL5;FCRH5;IRTA2;UNQ503/PRO820 | |
| IgG2a κ | |
| Protein G purified | |
| Clone F25 detects human Fc receptor-like protein 5. It targets an epitope with in the extracellular domain. |
| F25 | |
| ELISA, Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Inflammation & Immunology | |
| Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| NP_112571.2 | |
| Full-length FcRH5 expression plasmid followed by a boost with 293T cells transfected with FcRH5 plasmid. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title