Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRL1/FcRH1 Antibody (745916), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB374868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374868 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374868 Supplier Novus Biologicals Supplier No. FAB7286M
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FCRL1/FcRH1 Monoclonal specifically detects FCRL1/FcRH1 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen FCRL1/FcRH1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 745916
Conjugate DyLight 594
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD307a antigen, Fc receptor homolog 1, Fc receptor-like 1, FcRH1, FCRH1DKFZp667O1421, FcRL1, FcR-like protein 1, hIFGP1, IFGP family protein 1, IFGP1Fc receptor-like protein 1, Immune receptor translocation-associated protein 5, IRTA5immunoglobulin superfamily Fc receptor, gp42
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FCRL1/FcRH1, Ala17-Ser219, Accession # Q8R4Y0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115350
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.