Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRL2/FcRH2 Antibody (B24), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB151607 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151607 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151607 Supplier Novus Biologicals Supplier No. NBP311832JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCRL2/FcRH2 Monoclonal antibody specifically detects FCRL2/FcRH2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen FCRL2/FcRH2
Applications Flow Cytometry
Classification Monoclonal
Clone B24
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CD307b antigen, Fc receptor homolog 2, Fc receptor-like 2, FcRH2, FCRH2SPAP1B, FcRL2, FcR-like protein 2, IFGP family protein 4, IFGP4, Immunoglobulin receptor translocation-associated protein 4, immunoglobulin superfamily Fc receptor, gp42, IRTA4SPAP1C, SH2 domain containing phosphatase anchor protein 1, SH2 domain-containing phosphatase anchor protein 1Fc receptor-like protein 2, SPAP1A, SPAP1immune receptor translocation-associated protein 4
Host Species Mouse
Immunogen DNA-immunization followed by a boost with FCRL2/FcRH2 transfected cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79368
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.