Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRL4/FcRH4/IRTA1 Antibody (580810), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB365146 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365146 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365146 Supplier Novus Biologicals Supplier No. FAB24262J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCRL4/FcRH4/IRTA1 Monoclonal specifically detects FCRL4/FcRH4/IRTA1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen FCRL4/FcRH4/IRTA1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 580810
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD307d antigen, Fc receptor homolog 4, Fc receptor-like 4, FcRH4, FCRH4Fc receptor-like protein 4, FcRL4, FcR-like protein 4, hIFGP2, IFGP family protein 2, IFGP2, IGFP2, Immune receptor translocation-associated protein 1, immunoglobulin superfamily Fc receptor, gp42, immunoglobulin superfamily receptor translocation associated 1, IRTA1MGC150523, MGC150522
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human FCRL4/FcRH4, Gln16-Asp385, Accession # NP_112572
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83417
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.