Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRL5/FcRH5/IRTA2 Antibody (1010552), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB366482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366482 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366482 Supplier Novus Biologicals Supplier No. FAB20872X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCRL5/FcRH5/IRTA2 Monoclonal specifically detects FCRL5/FcRH5/IRTA2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen FCRL5/FcRH5/IRTA2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 1010552
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BXMAS1MGC119592, CD307, CD307e antigen, DKFZp667E2019, DKFZp667F216, Fc receptor homolog 5, Fc receptor-like 5, Fc receptor-like protein 5, FcRH5, FCRH5MGC119593, FcRL5, FcR-like protein 5, FLJ00333, FLJ00397, Immune receptor translocation-associated protein 2, immunoglobulin superfamily receptor translocation associated 2 (IRTA2), IRTA2MGC119590, PRO820
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human FCRL5 protein, Gln16-Arg844, Accession # Q96RD9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83416
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.