Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRL6/FcRH6 Antibody (672505) [DyLight 350], Novus Biologicals™
SDP

Catalog No. NB375863 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375863 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375863 Supplier Novus Biologicals Supplier No. FAB6745D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCRL6/FcRH6 Monoclonal antibody specifically detects FCRL6/FcRH6 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen FCRL6/FcRH6
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 672505
Conjugate DyLight 350
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Fc receptor-like 6, FLJ16056
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6 , Lys16-Leu312, Accession # Q6DN72
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343413
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.