Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRLA/FCRLM1 Antibody (865910) [PerCP], Novus Biologicals™
SDP

Catalog No. NB373917 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373917 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373917 Supplier Novus Biologicals Supplier No. FAB7905C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FCRLA/FCRLM1 Monoclonal specifically detects FCRLA/FCRLM1 in Mouse samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen FCRLA/FCRLM1
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 865910
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Fc receptor homolog expressed in B cells (FREB), Fc receptor homolog expressed in B-cells, Fc receptor related protein X, Fc receptor-like A, Fc receptor-like and mucin-like 1, Fc receptor-related protein X, FCRL1, FCRLa, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLFc receptor-like protein, FCRLM1, FCRLX, FCRX, FREBFc receptor-like and mucin-like protein 1, MGC4595
Host Species Rat
Immunogen E. coli-derived recombinant mouse FCRLA, Lys221-Lys352, Accession # Q920A9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84824
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.