Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRLB/FCRY Antibody (454217), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB347632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347632 Supplier Novus Biologicals Supplier No. FAB4705MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCRLB/FCRY Monoclonal antibody specifically detects FCRLB/FCRY in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen FCRLB/FCRY
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 454217
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FCRL2FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2FcRY, RP11-474I16.6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human FCRLB/FCRY, Ala18-Ser426, Accession # Q6BAA4
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127943
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.