Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRLB/FCRY Antibody (OTI5H6), HRP, Novus Biologicals™
SDP

Catalog No. NBP272384H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272384H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272384H Supplier Novus Biologicals Supplier No. NBP272384H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCRLB/FCRY Monoclonal specifically detects FCRLB/FCRY in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen FCRLB/FCRY
Applications Western Blot
Classification Monoclonal
Clone OTI5H6
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FCRL2FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2FcRY, RP11-474I16.6
Gene Symbols FCRLB
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 202-426 of human FCRLB/FCRY (NP_001002901) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 127943.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.